ContactUsLinkComponent
John Guest Speedfit Ball Valve, straight, 1/4" OD speedfit x 1/4" OD speedfit
John Guest Speedfit Ball Valve, straight, 1/4" OD speedfit x 1/4" OD speedfit
Catalog # MFLX07391-04
undefined
John Guest Speedfit Ball Valve, straight, 1/4" OD speedfit x 1/4" OD speedfit
Catalog # MFLX07391-04
About this item
Quickly and easily connect and disconnect valve.
  • Constructed of durable polypropylene (PP)
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.
  • Connection Type:
    1/4" OD Speedfit to 1/4" OD Speedfit
  • Max. pressure:
    10,3 bar (150 psi)
  • Max. temperature:
    65 °C (150 °F)
  • Description:
    Speedfit Ball Valve, straight, 1/4" OD speedfit x 1/4" OD speedfit
  • Environmentally Preferable:
  • Wetted materials:
    food-grade polypropylene, EPDM, and stainless steel
  • Warranty:
    1 year
  • Pk:
    1