Order Entry
Export
ContactUsLinkComponent
 
Micropump A-Mount Air-Driven Pump Drive, 500 to 8000 rpm, 5 to 11 scfm, 50 to 80 psi
  MFLX07144-97
undefined
Micropump A-Mount Air-Driven Pump Drive, 500 to 8000 rpm, 5 to 11 scfm, 50 to 80 psi
  MFLX07144-97

 

A variety of low-cost drives for numerous applications.

  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Hundreds of hours of dependable leakproof service

These Economical Drives for Micropump A-Mount Pump Heads are designed with magnetic coupling to minimize friction. Choose from AC motor drives, DC motor drives, explosion-proof drives, or air-powered drives.

Explosion-proof and air-powered models are ideal for safe pumping in hazardous areas. Explosion-proof drives are rated for NEMA Class 1, Groups C and D locations.

  • Power:
    115/230 VAC; Air
  • Description:
    A-Mount Air-Driven Pump Drive, 500 to 8000 rpm, 5 to 11 ft.³/min, 50 to 80 psi
  • Height:
    133 mm
  • Length:
    9,5 cm
  • Environmentally Preferable:
  • Duty cycle continuous:
    Yes
  • Max. air inlet pressure:
    5,5 bar (80 psig)
  • Min. motor RPM:
    500 rpm
  • Motor enclosure type:
    TENV
  • Motor hp:
    0,13 to 0,29
  • Air consumption:
    18 m³/h (11 ft.³/min)
  • Min. temperature:
    4 °C (40 °F)
  • Max. temperature:
    60 °C (140 °F)
  • Warranty:
    1 year
  • Pk:
    1
  • Width:
    7,6 cm
  • Max. motor RPM:
    8000 rpm