Micropump Economical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC
Catalog # MFLX07003-90
Supplier: Avantor Fluid Handling
About this item
A variety of low-cost drives for numerous applications.
- Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
- Hundreds of hours of dependable leakproof service
These Economical Drives for Micropump A-Mount Pump Heads are designed with magnetic coupling to minimize friction. Choose from AC motor drives, DC motor drives, explosion-proof drives, or air-powered drives.
Explosion-proof and air-powered models are ideal for safe pumping in hazardous areas. Explosion-proof drives are rated for NEMA Class 1, Groups C and D locations.
- Power:0 - 24 VDC; 3 A
- Description:Economical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC
- Height:76 mm
- Length:10,2 cm
- Environmentally Preferable:
- Duty cycle continuous:Yes
- Min. motor RPM:300 rpm
- Motor enclosure type:PM brush, TENV
- Motor hp:0,09
- Min. temperature:4 °C (40 °F)
- Max. temperature:60 °C (140 °F)
- Warranty:1 year
- Pk:1
- Width:7,6 cm
- Max. motor RPM:4000 rpm