Orders LinkContactUsLinkComponent
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Easy-Load® 3 Pump Head Mounting Hardware, Mounts One Pump Head, Stainless steel, Masterflex L/S® Easy-Load® 3 Pump Head, L/S
Catalog # MFLX77800-00
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Catalog # MFLX77800-00
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Easy-Load® 3 Pump Head Mounting Hardware, Mounts One Pump Head, Stainless steel, Masterflex L/S® Easy-Load® 3 Pump Head, L/S
Price
77,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
  • Constructed of durable stainless steel
  • See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Documents
Specifications
  • Description:
    L/S® Easy-Load® 3 Pump Head Mounting Hardware, Mounts One Pump Head
  • Material:
    Stainless steel
  • Seq:
    11
  • Compatible with:
    Masterflex L/S® Easy-Load® 3 Pump Head
  • Pump series:
    L/S