Cleanroom ESD Ergo Chairs
Cleanroom chair with conductive locking casters
Catalog # PALBPCKD0110
Supplier: Palbam
Jump to:
Product Details & Documents
Cleanroom ESD ergo chair which lives up to our reputation for high quality and ergonomics/operator comfort.
- Seat and backrest material and colour: Polyurethane (PU), Black (RAL 9011)
- Base Material: Chrome steel
Seat height adjustable from 450 to 640 mm.
The chairs include the 'automatic weight adjustment' ergo feature and a tapering backrest offering optimum freedom for shoulders. Options available for conductive glides or casters.
Certifications: The chairs comply with: Cleanliness class 4 DIN EN ISO 14644-1 Anti-static and conductive according to DIN EN 61340-5-1:2000.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Description:Cleanroom chair with conductive locking casters
- Seq:02