Order Entry
Export
Orders LinkContactUsLinkComponent
Defenders® Indoor Step-On Containers

Defenders® Indoor Step-On Containers

Stainless steel, Metal, 95 L, 480×480×783 mm
Catalog # 129-1952
Defenders® Indoor Step-On Containers
Catalog # 129-1952
Defenders® Indoor Step-On Containers
Defenders® Indoor Step-On Containers

Stainless steel, Metal, 95 L, 480×480×783 mm

Price
1 170,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Metal step-on containers safely and hygienically collect waste.

  • Step-on foot pedal enables hands-free operation, optimising sanitary efforts
  • Smooth surfaces are easy to clean
  • Heavy-gauge, fire-safe steel for years of reliable service
  • Restraint mechanism and non-slip feet protect walls and floors
  • Rigid plastic liner included to contain liquids and keep the surrounding area clean
  • Includes a 5-year warranty against normal wear and tear

The Defenders® indoor step-On container is an ideal waste container for hospitals, doctor’s offices, and other healthcare facilities. The lid has a restraint mechanism to protect walls from scuffs, as well as a vinyl base ring to protect floors.

Certifications: Constructed of fire-safe galvanised steel for CSFM and UL certifications. Complies with OSHA standards for containment of infectious waste. This fire-safe steel receptacle is FM certified.

Specifications
  • Capacity:
    95 L
  • Colour:
    Stainless steel
  • Material:
    Metal
  • Seq:
    05
  • W×D×H:
    480×480×783 mm