Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
ARMAND STAINING SOLUTION 1L 1 * 1 L
ARMAND STAINING SOLUTION 1L 1 * 1 L
Print
ARMAND STAINING SOLUTION 1L 1 * 1 L
Catalog # 720-0339
Supplier:
RAL
ARMAND STAINING SOLUTION 1L 1 * 1 L
Catalog # 720-0339
Supplier:
RAL
CAS Number:
Expand image
Return to Top
ARMAND STAINING SOLUTION 1L 1 * 1 L
Add to cart
CAS Number:
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 L
-
+
Inquire For Price
Return Policy
Product Details & Documents