MicroCNX™ connectors
MicroCNX™ connector, hose barb size, 2,4 mm
212-0792
Aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small format biomanufacturing assemblies. MicroCNX™ connectors are the modern alternative to the cumbersome, industrial process of tube welding.
- Pinch – click and pull: Intuitive three-step connection process reduces risk of operator error
- Easy to use: Lowers risk of operator error and related performance, reliability and safety concerns
- Genderless: Eases single-use systems specifications with one part number for both halves
- CPC click: Audible confirmation of assembly with no additional hardware required
The MicroCNX™ line is engineered specifically for the challenging conditions of biologic media transfer in bioprocessing, cell therapy and gene therapy applications.
Autoclave: One cycle up to 130 °C (266 °F) for 60 minutes
Sterilisation: Up to 50 kGy gamma irradiation
Main components: Polycarbonate (white); USP Class VI, ADCF
Seals: Silicone (clear), platinum-cured; USP Class VI, ADCF
Protective cover: Polypropylene (teal); USP Class VI, ADCF
Membrane: Hydrophobic polyethersulphone; USP Class VI
- Description:MicroCNX™ connector, hose barb size, 2,4 mm
- Seq:02
- Cat. No.:212-0792
- Max. temperature:1...40 °C
- Max. operating pressure:Up to 4,1 bar; Up to 5,1 bar for 48 hours



