Order Entry
Export
Orders LinkContactUsLinkComponent
MicroCNX™ connectors

MicroCNX™ connectors

MicroCNX™ connector, hose barb size, 3,2 mm
Catalog # 212-0791
Supplier:   CPC COLDER
MicroCNX™ connectors
Catalog # 212-0791
Supplier:  CPC COLDER
MicroCNX™ connectors
MicroCNX™ connectors

MicroCNX™ connector, hose barb size, 3,2 mm

Price
794,00 €
Log in for availability
1 * 25 items
Product Details & Documents

Aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small format biomanufacturing assemblies. MicroCNX™ connectors are the modern alternative to the cumbersome, industrial process of tube welding.

  • Pinch – click and pull: Intuitive three-step connection process reduces risk of operator error
  • Easy to use: Lowers risk of operator error and related performance, reliability and safety concerns
  • Genderless: Eases single-use systems specifications with one part number for both halves
  • CPC click: Audible confirmation of assembly with no additional hardware required

The MicroCNX™ line is engineered specifically for the challenging conditions of biologic media transfer in bioprocessing, cell therapy and gene therapy applications.

Autoclave: One cycle up to 130 °C (266 °F) for 60 minutes
Sterilisation: Up to 50 kGy gamma irradiation

Main components: Polycarbonate (white); USP Class VI, ADCF
Seals: Silicone (clear), platinum-cured; USP Class VI, ADCF
Protective cover: Polypropylene (teal); USP Class VI, ADCF
Membrane: Hydrophobic polyethersulphone; USP Class VI

Specifications
  • Description:
    MicroCNX™ connector, hose barb size, 3,2 mm
  • Seq:
    03
  • Cat. No.:
    212-0791
  • Max. temperature:
    1...40 °C
  • Max. operating pressure:
    Up to 4,1 bar; Up to 5,1 bar for 48 hours