Order Entry
Export
Orders LinkContactUsLinkComponent
Grounding cables, b.safe

Grounding cables, b.safe

Grounding cable with clip and 13 - 18 mm Ø tube clip, 3,0 m
Catalog # BOHLR606-05
Supplier:   Bohlender
Grounding cables, b.safe
Catalog # BOHLR606-05
Supplier:  Bohlender
Grounding cables, b.safe
Grounding cables, b.safe

Grounding cable with clip and 13 - 18 mm Ø tube clip, 3,0 m

Price
32,00 €
Log in for availability
1 * 1 items
Product Details & Documents

For earth connection of vessels and components made of static dissipative materials.

  • Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
  • Customised versions can be offered on request
  • Suitable grounding accessories for b.safe products

The scope of delivery of the ring cable lug includes screw, nut and washer.

Specifications
  • Description:
    Grounding cable with clip and 13 - 18 mm Ø tube clip
  • Length:
    3,0 m
  • Seq:
    10