Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings, BLAUBRAND®
Volumetric flasks, class A, blue markings, BLAUBRAND®
150 ml, 14/23, 0,15 ± ml
Catalog # 612-7122
Supplier:   Brand
Volumetric flasks, class A, blue markings, BLAUBRAND®
Catalog # 612-7122
Supplier:  Brand
Volumetric flasks, class A, blue markings, BLAUBRAND®
Volumetric flasks, class A, blue markings, BLAUBRAND®
150 ml, 14/23, 0,15 ± ml
Price
54,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Individual certificate included
* In addition to DIN.
**Wide neck.
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    14/23
  • Capacity:
    150 ml
  • Seq:
    12
  • Tolerance:
    0,15 ± ml