Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Reprospher Column 100 C30-DE, 3 µm, 150mmX4.6mm 1 * 1 items
Catalog # MAISRS13.3DE.S1546
Supplier:
DR MAISCH
Expand image
Reprospher Column 100 C30-DE, 3 µm, 150mmX4.6mm 1 * 1 items
Catalog # MAISRS13.3DE.S1546
Supplier:
DR MAISCH