Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
pH 9.000 MILLESIMAL BUFFER SOLUTION (±0. 1 * 500 mL
pH 9.000 MILLESIMAL BUFFER SOLUTION (±0. 1 * 500 mL
Print
pH 9.000 MILLESIMAL BUFFER SOLUTION (±0. 1 * 500 mL
Catalog # HANNHI-6009
Supplier:
Hanna
pH 9.000 MILLESIMAL BUFFER SOLUTION (±0. 1 * 500 mL
Catalog # HANNHI-6009
Supplier:
Hanna
Expand image
Return to Top
pH 9.000 MILLESIMAL BUFFER SOLUTION (±0. 1 * 500 mL
Add to cart
Price
72,60 €
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Return Policy
Product Details & Documents