Potassium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
2,5 kg
Supplier: Avantor
Potassium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Catalog # 3152-35
Supplier: Avantor
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (1013 hPa)
- Melting Pt:360 °C
- Density:2.04 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1813
- ADR:8,II
- REACH:01-2119487136-33
Documents
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Insoluble Substances:Passes Test
- NF - Content of Potassium:≥59.9 %
- NF - Limit of Sodium:≤1.0 %
- NF - Assay (Total Alkali):≥85.0 %
- NF - Carbonate (as K₂CO₃):≤3.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Carbonate (as K₂CO₃):≤2.0 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Phosphate (PO₄):≤100 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Sodium (Na):≤1.0 %
- EP/BP - Assay:85.0 - 100.5 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.050 %
- JP - Heavy Metals (as Pb):≤30 ppm
- JP - Sodium (Na):Passes Test
- JP - Potassium Carbonate (K₂CO₃):≤2.0 %
- JP - Assay (KOH):≥85.0 %
- No Class 1,2,3 or other solvents are used or produced in the:= 85.0 %
Specifications
- Pk:2,5 kg



