Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SNAP CAP VIAL 20ML WHITE GLASS SAFE PACK (31.0Ø mm, 51.0 Höhe mm ,27.0 Ø Deckel mm) 1 * 210 items
Catalog # MUEL14.021.34
Supplier:
MUELLER KREMPEL
Expand image
SNAP CAP VIAL 20ML WHITE GLASS SAFE PACK (31.0Ø mm, 51.0 Höhe mm ,27.0 Ø Deckel mm) 1 * 210 items
Catalog # MUEL14.021.34
Supplier:
MUELLER KREMPEL