Orders LinkContactUsLinkComponent
Wash troughs
Knee operated five station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 2505W×380D×1250H mm, 2500×123×283 mm
  TEKMWTN050EXPED
 :   TEKNOMEK
Wash troughs
  TEKMWTN050EXPED
 :  TEKNOMEK
Wash troughs
Wash troughs
Knee operated five station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 2505W×380D×1250H mm, 2500×123×283 mm
1 * 1 items
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienic standards.
 : Optional skirt (to be ordered alongside the wash trough as cannot be retrofitted).
  • Description:
    Knee operated five station wash trough, pedestal mounted, fully welded 304-grade stainless steel, 2505W×380D×1250H mm
  • Seq:
    27
  • W×D×H int.:
    2500×123×283 mm