Orders LinkContactUsLinkComponent
Wash troughs
Wash troughs
Knee operated three station wash trough, wall mounted, fully welded 304-grade stainless steel, 1500W×380D×525H mm, 1500×123×283 mm
Catalog # TEKMWTN030EX
Supplier:   TEKNOMEK
Wash troughs
Catalog # TEKMWTN030EX
Supplier:  TEKNOMEK
Wash troughs
Wash troughs
Knee operated three station wash trough, wall mounted, fully welded 304-grade stainless steel, 1500W×380D×525H mm, 1500×123×283 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienic standards.
Ordering information: Optional skirt (to be ordered alongside the wash trough as cannot be retrofitted).
Specifications
  • Description:
    Knee operated three station wash trough, wall mounted, fully welded 304-grade stainless steel, 1500W×380D×525H mm
  • Seq:
    13
  • W×D×H int.:
    1500×123×283 mm