Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top
Computer desk, solid top, 1800×750×840 mm
TEKMTSSC1800X750
: TEKNOMEK
Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top
TEKMTSSC1800X750
: TEKNOMEK
Stainless steel computer desk wih pull out keyboard and support for PC tower, available in solid or perforated top for optimum air flow.
- Computer desk with top and tower shelf - silicone seal
- Fully welded 304 grade stainless steel, no construction or assembly required
- Brushed dull polish finish
- Solid or perforated top
- Keyboard/mouse drawer
- Stainless steel adjustable feet with non-marking rubber base
A ledge for a regular size computer tower and a keyboard shelf, also manufactured from 304 grade stainless steel, this desk is designed to house your computer safely.
Load capaciity: 250 kg
- Description:Computer desk, solid top
- Seq:10
- W×D×H:1800×750×840 mm



