Order Entry
Export
Orders LinkContactUsLinkComponent
Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top

Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top

Computer desk, solid top, 1500×750×840 mm
Catalog # TEKMTSSC1500X750
Supplier:   TEKNOMEK
Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top
Catalog # TEKMTSSC1500X750
Supplier:  TEKNOMEK
Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top
Cleanroom computer desks for PC with pull-out keyboard, perforated or solid top

Computer desk, solid top, 1500×750×840 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel computer desk wih pull out keyboard and support for PC tower, available in solid or perforated top for optimum air flow.

  • Computer desk with top and tower shelf - silicone seal
  • Fully welded 304 grade stainless steel, no construction or assembly required
  • Brushed dull polish finish
  • Solid or perforated top
  • Keyboard/mouse drawer
  • Stainless steel adjustable feet with non-marking rubber base

A ledge for a regular size computer tower and a keyboard shelf, also manufactured from 304 grade stainless steel, this desk is designed to house your computer safely.

Load capaciity: 250 kg

Specifications
  • Description:
    Computer desk, solid top
  • Seq:
    08
  • W×D×H:
    1500×750×840 mm