Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dollies

Mobile roller bogie, stainless steel for 600×400 mm tray, 600×400×175 mm
Catalog # TEKMBS100006040S
Supplier:   TEKNOMEK
Dollies
Catalog # TEKMBS100006040S
Supplier:  TEKNOMEK
Dollies
Dollies

Mobile roller bogie, stainless steel for 600×400 mm tray, 600×400×175 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Our stainless steel circular dollies are a extremely robust, with an up to 150 kg payload.

  • Fully welded 304 grade stainless steel
  • Brushed dull polish finish
  • 4 swivel castors
  • Crevice free
  • Sturdy design
  • Easy-clean design
  • Fully welded, ready to use

They come with swivel castors, ensuring smooth and effortless movement. Rectangular stainless steel and aluminum bogies are also available.

Specifications
  • Description:
    Mobile roller bogie, stainless steel for 600×400 mm tray
  • Seq:
    06
  • W×D×H:
    600×400×175 mm