Dollies
Circular dollie, 600 mm Ø stainless steel, 600×600×165 mm
TEKMBS0010/600
: TEKNOMEK
Our stainless steel circular dollies are a extremely robust, with an up to 150 kg payload.
- Fully welded 304 grade stainless steel
- Brushed dull polish finish
- 4 swivel castors
- Crevice free
- Sturdy design
- Easy-clean design
- Fully welded, ready to use
They come with swivel castors, ensuring smooth and effortless movement. Rectangular stainless steel and aluminum bogies are also available.
- Description:Circular dollie, 600 mm Ø stainless steel
- Seq:05
- W×D×H:600×600×165 mm



