Orders LinkContactUsLinkComponent
Modaflame Multi Norm Arc Softshell Jackets
Modaflame Multi Norm Arc Softshell Jackets
Orange/dark blue, M, Regular
Catalog # PRTWMV72ONRM
Supplier:   Portwest
Modaflame Multi Norm Arc Softshell Jackets
Catalog # PRTWMV72ONRM
Supplier:  Portwest
Modaflame Multi Norm Arc Softshell Jackets
Orange/dark blue, M, Regular
Price
253,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The anti-static Modaflame multi norm arc softshell jacket is constructed from a highly protective three-layer softshell fabric. Using modacrylic and carbon fibre elements, the softshell provides flame, chemical, anti-static, and high visibility protection. An interactive zipper allows this softshell to be zipped into an outer jacket for additional insulation if required.
  • Durable, breathable, windproof and water-resistant fabric
  • Inherent flame resistant qualities will not diminish with washing
  • Protects against radiant and convective heat
  • Wind-resistant to protect against wind chill
  • Hook and loop cuffs for a secure fit
  • Interactive zip for added functionality
  • Premium sew-on flame-resistant reflective tape; contrast colouring for added style
The Modaflame multi norm arc softshell jacket features include a drawcord adjustable hem, storm flap front to protect against the elements, a chin guard for added comfort and stability, a radio loop for easy clipping of a radio, flame-resistant zip, and zipped pockets.
Certifications: EN ISO 11612 A1, B1, C2. EN ISO 11611 Class 2 A1. EN 1149 -5. IEC 61482-2 IEC 61482-1-2 APC 2. IEC 61482-2 IEC 61482-1-1 ELIM 11 cal/cm². EN 13034 Type PB [6]. EN ISO 20471 Class 3. RIS 3279 TOM Issue 2 (orange only).
Specifications
  • Clothing Size:
    M
  • Garment fit:
    Regular
  • Colour:
    Orange/dark blue
  • Seq:
    02
  • Cat. No.:
    PRTWMV72ONRM
  • Sleeve or leg length:
    Long
  • Risk area:
    Flame-retardant
  • Garment application:
    Fire protection
  • Gender:
    For men
  • Closure type:
    Zip closure
  • Fabric:
    100% polyester, laminated to TPU FR membrane, bonded with 60% modacrylic, 39% cotton, 1% carbon fibre