Hand wash, SURE®
SURE® Hand Wash, 0,5 L
Catalog # 115-2954
Supplier: Diversey
Jump to:
Product Details & Documents
Gentle and effective hand wash with a 100% natural perfume. Cleans hands leaving them hygienically fresh.
- Free from parabens and other preservatives.
- Contains plant-based actives from renewable sources
- Decomposed by natural processes
- No environmental hazard classifications, no safety warnings for product or user
- 100% natural perfume with a refreshing lemon fragrance
- Skin compatability tested (Patch test)
SURE® is a comprehensive offering of plant-based, cleaning products designed to deliver superior professional results while being safe for people and kind to the environment.
With environmentally-friendly ingredients from renewable resources, SURE® products meet the daily requirements for kitchens, facilities, and personal care.
Certifications: OECD 301B. Cradle to cradle certification at gold level.
Ordering information: 1.3 L cartridge is suitable for Diversey IntelliCare Dispenser (manual/hybrid).
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
TDS Diversey SURE Hand Wash
Datasheet
Specifications
- Description:SURE® Hand Wash, 0,5 L
- Seq:01



