Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Furnace chamber lf60/14 b400, tmax. 1400 1 * 1 items
Catalog # 455-2175
Supplier:
NABERTHERM
Expand image
Furnace chamber lf60/14 b400, tmax. 1400 1 * 1 items
Catalog # 455-2175
Supplier:
NABERTHERM