Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W3) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 7 - Thr 1129 (ITGA2) & Gln 21 - Asp 728 (ITGB1) (Accession # Q62469-1 (ITGA2) & P09055-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-M52W3100UG
Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W3) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 7 - Thr 1129 (ITGA2) & Gln 21 - Asp 728 (ITGB1) (Accession # Q62469-1 (ITGA2) & P09055-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-M52W3100UG