Orders LinkContactUsLinkComponent
Accessories for multi-use data loggers, tempmate.®-M2
Accessories for multi-use data loggers, tempmate.®-M2
tempmate.®-M2 External digital PT100 temperature sensor; 1,2 m length, –80...+200 °C
Catalog # IMECM2S-300
Supplier:   tempmate
Accessories for multi-use data loggers, tempmate.®-M2
Catalog # IMECM2S-300
Supplier:  tempmate
Accessories for multi-use data loggers, tempmate.®-M2
Accessories for multi-use data loggers, tempmate.®-M2
tempmate.®-M2 External digital PT100 temperature sensor; 1,2 m length, –80...+200 °C
Price
68,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    tempmate.®-M2 External digital PT100 temperature sensor; 1,2 m length, –80...+200 °C
  • Seq:
    01