Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Perforated basket, O 320 mm, height 250 mm,for Autoclaves WAC-60 1 * 1 items
WITGDH.WAC20060
:
witeg Labortechnik
Perforated basket, O 320 mm, height 250 mm,for Autoclaves WAC-60 1 * 1 items
WITGDH.WAC20060
:
witeg Labortechnik