Order Entry
Export
Orders LinkContactUsLinkComponent
 

iFSP1 ≥98%

100 mg
Catalog # CAYM29483-100
Irritant
Supplier:   Cayman Chemical
iFSP1 ≥98%
Catalog # CAYM29483-100
Supplier:  Cayman Chemical
CAS Number:  150651-39-1
iFSP1 ≥98%

100 mg

CAS Number:  150651-39-1
Price
684,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

iFSP1 is an inhibitor of apoptosis inducing factor mitochondria-associated 2/ferroptosis suppressor protein 1 (AIFM2/FSP1)

 : 98

Synonyms:  1-amino-3-(4-methylphenyl)-pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile
  • CAS Number:150651-39-1
  • Formula:C₂₀H₁₃N₅
  • MDL Number:MFCD01572665
  • Molecular Weight:323.4 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    100 mg

Specifications