Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Aluminium soil sample box, cont. 120 cc Aluminium soil sample box with lid, Ø 78 x 27 mm, content 120 cc 1 * 1 ST
Catalog # EIJK99080303
Supplier:
EIJKELKAMP AGRISECTOR
Expand image
Aluminium soil sample box, cont. 120 cc Aluminium soil sample box with lid, Ø 78 x 27 mm, content 120 cc 1 * 1 ST
Catalog # EIJK99080303
Supplier:
EIJKELKAMP AGRISECTOR