VWR® Deep Well Plates, Square Well
Deep well plate, 96 square well, V-bottom, Clear, 5 per inner pack, -, 1,0 ml
Supplier: VWR Collection
Jump to:
Product Details & Documents
These deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
Chemical resistant
- Free from DNase and RNase
- V-bottom well shape maximises liquid removal and aids resuspension
- U bottom suited for washing or stirring samples
- No inner edges to interfere with magnetic bead collection
- Raised well rims to improve heat sealing
-
Conical base aids sample concentration, reconstitution and centrifugation
Suitable for use with chromatographic applications
Minimum order quantity of 40 cases for barcoded plates.
: Can be autoclaved once, but only if not used with robotic systems.
Specifications
- Packed:5 per inner pack
- Sterile:-
- Colour:Clear
- Description:Deep well plate, 96 square well, V-bottom
- Seq:01
- Well volume:1,0 ml
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cap mat applicator
Product Information
- Catalog #AvailabilityPrice
- Cap mat, 96 square well, EVA
Product Information
- Catalog #AvailabilityPrice
- Cap mat, 96 square well, PP