Order Entry
Export
Orders LinkContactUsLinkComponent
epT.I.P.S.® Box 2.0 Pipette Tips

epT.I.P.S.® Box 2.0 Pipette Tips

0,1 - 20 µl, -, 40 mm, Grey, 1 reusable box × 96 tips
Catalog # EPPE0030076.133
   Low Manufacturing Impact
Supplier:   EPPENDORF
epT.I.P.S.® Box 2.0 Pipette Tips
Catalog # EPPE0030076.133
Supplier:  EPPENDORF
   Low Manufacturing Impact
epT.I.P.S.® Box 2.0 Pipette Tips
epT.I.P.S.® Box 2.0 Pipette Tips

0,1 - 20 µl, -, 40 mm, Grey, 1 reusable box × 96 tips

Price
43,40 €
Log in for availability
1 * 96 items
Product Details & Documents

Eppendorf epT.I.P.S.® Box 2.0, available with 'Eppendorf Quality' pipette tips or as an empty box, improves lab efficiency with innovative TwinLid® technology, enhanced stackability, and superior ergonomics for safe and organised pipetting.

  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121 °C for subsequent use
  • epT.I.P.S.® Box 2.0 and epT.I.P.S.® Set available in Eppendorf Quality
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally Integrated Pipetting System') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognise an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0,5 to 20 µl, 50 to 1250 µl, 0,2 to 5 ml, and 0,5 to 10 ml.

Certifications: General quality and ISO certificates available.

Specifications
  • Colour code:
    Grey
  • Packed:
    1 reusable box × 96 tips
  • Sterile:
    -
  • Volume:
    0,1 - 20 µl
  • Length:
    40 mm
  • Seq:
    02
  • Cat. No.:
    EPPE0030076.133
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact