Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Metall tubing triple insulated (-100°C...+350°C), 2.0 m Metal tubing with connector M16x1 1 * 2 m
Catalog # JULA8930216
Supplier:
JULABO GmbH
Expand image
Metall tubing triple insulated (-100°C...+350°C), 2.0 m Metal tubing with connector M16x1 1 * 2 m
Catalog # JULA8930216
Supplier:
JULABO GmbH