VWR®, Transfer Pipettes, Graduated
1 ml (max. 5 ml), 23, 3 ml, 0,25 - 1 ml, +, Individually packed
Supplier: VWR Collection
Jump to:
Product Details & Documents
These LDPE disposable, unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
- Will not shatter or break
- Non toxic and inert to biological fluids and most acids
- Can be sealed, refrigerated and gas sterilised
- No bulb to insert or remove
- Uniform drop size
They work well whenever there is a need for quick, safe transfer of fluids.
: Compliance to US FDA 21 CFR.
Specifications
- Packed:Individually packed
- Sterile:+
- Capacity:1 ml (max. 5 ml)
- Seq:02
- Drops per ml:23
- Graduation:0,25 - 1 ml
- Bulb draw:3 ml



