Orders LinkContactUsLinkComponent
Accessories for Stuart Temperature Controller SCT2
Accessories for Stuart Temperature Controller SCT2
Stuart® temperature probe for temperature controller, PTFE, temperature range 50...200 °C with ±0,5 °C accuracy
Catalog # STUA04805-93
Supplier:   Cole Parmer
Accessories for Stuart Temperature Controller SCT2
Catalog # STUA04805-93
Supplier:  Cole Parmer
Accessories for Stuart Temperature Controller SCT2
Accessories for Stuart Temperature Controller SCT2
Stuart® temperature probe for temperature controller, PTFE, temperature range 50...200 °C with ±0,5 °C accuracy
Price
161,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Stuart® temperature probe for temperature controller, PTFE, temperature range 50...200 °C with ±0,5 °C accuracy
  • Seq:
    02