Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STREAKING HEAD 8 PINS 3.0MM DIAMETER 1 * 1 items
Catalog # MLDVX4345
Supplier:
Molecular Devices
Expand image
STREAKING HEAD 8 PINS 3.0MM DIAMETER 1 * 1 items
Catalog # MLDVX4345
Supplier:
Molecular Devices