Uric acid
Bottle, 100 g
Catalog # APOSBIK9021-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 1H-Purine-2,6,8(3H,7H,9H)-trione
- CAS Number:69-93-2
- Formula:C₅H₄N₄O₃
- MDL Number:MFCD00005712
- Molecular Weight:168.11 g/mol
- Melting Pt:>300 °C
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:200-720-7
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



