Order Entry
Export
Orders LinkContactUsLinkComponent
 

Quercetin, anhydrous

Bottle, 25 g
Catalog # APOSBIK9011-25G
Toxic
Supplier:   Apollo Scientific
Quercetin, anhydrous
Catalog # APOSBIK9011-25G
Supplier:  Apollo Scientific
CAS Number:  117-39-5
Quercetin, anhydrous

Bottle, 25 g

CAS Number:  117-39-5
Price
34,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-5-7-3-4-Pentahydroxyflavone, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,5,7,3',4'-Pentahydroxyflavone
  • CAS Number:117-39-5
  • Formula:C₁₅H₁₀O₇
  • MDL Number:MFCD00149487
  • Molecular Weight:302.25 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-187-1
  • UN:2811
  • ADR:6.1,III

 

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications