Order Entry
Export
Orders LinkContactUsLinkComponent
 

Calcein

Bottle, 5 g
Catalog # APOSBIB1007-5G
Supplier:   Apollo Scientific
Calcein
Catalog # APOSBIB1007-5G
Supplier:  Apollo Scientific
CAS Number:  154071-48-4
Calcein

Bottle, 5 g

CAS Number:  154071-48-4
Price
138,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
  • CAS Number:154071-48-4
  • Formula:C₃₀H₂₆N₂O₁₃
  • MDL Number:MFCD00005049
  • Molecular Weight:622.53 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications