Order Entry
Export
Orders LinkContactUsLinkComponent
Hydroxy-PEG2-CH2CO2t-Bu

Hydroxy-PEG2-CH2CO2t-Bu

Bottle, 1 g
Catalog # APOSBIPG1436-1G
Irritant
Supplier:   Apollo Scientific
Hydroxy-PEG2-CH2CO2t-Bu
Catalog # APOSBIPG1436-1G
Supplier:  Apollo Scientific
CAS Number:  149299-82-1
Hydroxy-PEG2-CH2CO2t-Bu

Bottle, 1 g

CAS Number:  149299-82-1
Price
333,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  tert-Butyl 2-(2-(2-hydroxyethoxy)ethoxy)acetate, Hydroxy-PEG2-CH2-Boc
  • CAS Number:149299-82-1
  • Formula:C₁₀H₂₀O₅
  • MDL Number:MFCD25959245
  • Molecular Weight:220.263 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications