Order Entry
Export
Orders LinkContactUsLinkComponent
Biotin-PEG4-amine

Biotin-PEG4-amine

Bottle, 50 mg
Catalog # APOSBIPG1209-50MG
Irritant
Supplier:   Apollo Scientific
Biotin-PEG4-amine
Catalog # APOSBIPG1209-50MG
Supplier:  Apollo Scientific
CAS Number:  663171-32-2
Biotin-PEG4-amine

Bottle, 50 mg

CAS Number:  663171-32-2
Price
138,00 €
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  N-(14-Amino-3,6,9,12-tetraoxatetradecyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Amine-PEG4-Biotin, N-Biotinyl-3,6,9,12-tetraoxatetradecane-1,14-diamine, N-[2-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide
  • CAS Number:663171-32-2
  • Formula:C₂₀H₃₈N₄O₆S
  • MDL Number:MFCD26142992
  • Molecular Weight:462.606 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg

Specifications