Order Entry
Export
Orders LinkContactUsLinkComponent
17-DMAG

17-DMAG

Bottle, 25 mg
Catalog # APOSBID1002-25MG
Irritant
Supplier:   Apollo Scientific
17-DMAG
Catalog # APOSBID1002-25MG
Supplier:  Apollo Scientific
CAS Number:  467214-20-6
17-DMAG

Bottle, 25 mg

CAS Number:  467214-20-6
Price
218,00 €
Log in for availability
1 * 25 mg
Product Details & Documents
Synonyms:  17-Dimethylaminoethylamino-17-demethoxygeldanamycin
  • CAS Number:467214-20-6
  • Formula:C₃₂H₄₈N₄O₈
  • MDL Number:MFCD08457919
  • Molecular Weight:616.8 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg

Specifications