Order Entry
Export
Orders LinkContactUsLinkComponent
J.T.Baker®, Plate 96

J.T.Baker®, Plate 96

PP plates, U-bottom, Transparent, -, 0,3 ml
Catalog # 43001-0600
   Low Manufacturing Impact
Supplier:   Avantor
J.T.Baker®, Plate 96
Catalog # 43001-0600
Supplier:  Avantor
   Low Manufacturing Impact
J.T.Baker®, Plate 96
J.T.Baker®, Plate 96

PP plates, U-bottom, Transparent, -, 0,3 ml

Price
68,09 €
Log in for availability
1 * 64 items
Product Details & Documents

J.T.Baker Plate 96 round well plates made from PS are ideal for ELISA applications.

  • Alphanumeric coding assists with identification of samples
  • Plate shape significantly reduces sample carry-over
  • Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
Specifications
  • Sterile:
    -
  • Colour:
    Transparent
  • Description:
    PP plates, U-bottom
  • Well volume:
    0,3 ml
  • Cat. No.:
    43001-0600
  • Environmental certification:
    ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU
  • Environmentally Preferable:
    Low Manufacturing Impact