Order Entry
Export
Orders LinkContactUsLinkComponent
J.T.Baker®, Plate 96

J.T.Baker®, Plate 96

PS plates, V-bottom, Clear, -, 0,24 ml
Catalog # 43001-0118
   Low Manufacturing Impact
Supplier:   Avantor
J.T.Baker®, Plate 96
Catalog # 43001-0118
Supplier:  Avantor
   Low Manufacturing Impact
J.T.Baker®, Plate 96
J.T.Baker®, Plate 96

PS plates, V-bottom, Clear, -, 0,24 ml

Price
106,09 €
Log in for availability
1 * 100 items
Product Details & Documents

J.T.Baker Plate 96 round well plates made from PS are ideal for ELISA applications.

  • Alphanumeric coding assists with identification of samples
  • Plate shape significantly reduces sample carry-over
  • Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
Specifications
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    PS plates, V-bottom
  • Well volume:
    0,24 ml
  • Cat. No.:
    43001-0118
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Environmentally Preferable:
    Low Manufacturing Impact