J.T.Baker®, Plates, Square Well
24-well plate, Transparent, -, 10,0 ml
Catalog # 43001-0066
Supplier: Avantor
Jump to:
Product Details & Documents
These deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
- Chemical resistant
- V-bottom shape for 24- and 48-wells maximises liquid removal and aids resuspension
- U-bottom shape for 96 wells
- No inner edges to interfere with magnetic bead collection
- Raised well rims to improve heat sealing
- Conical base aids sample concentration, reconstitution and centrifugation
For a maximum capacity utilisation.
Specifications
- Sterile:-
- Colour:Transparent
- Description:24-well plate
- Well volume:10,0 ml
- Cat. No.:43001-0066
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #Optical propertyMaterialAvailabilityPrice
- Capmat for J.T.Baker plate square well 96Capmat for J.T.Baker plate square well 9643001-0022TransparentEVALog in for availability1 * 20 items - 49,13 €
Product Information
- Catalog #Optical propertyMaterialAvailabilityPrice
- Capmat for J.T.Baker plate square well 48Capmat for J.T.Baker plate square well 4843001-0069TransparentEVALog in for availability1 * 20 items - 57,88 €
Product Information
- Catalog #Optical propertyMaterialAvailabilityPrice
- Capmat for J.T.Baker plate square well 24Capmat for J.T.Baker plate square well 2443001-0070TransparentEVALog in for availability1 * 20 items - 63,65 €