Orders LinkContactUsLinkComponent
J.T.Baker®, Plates, Square Well
J.T.Baker®, Plates, Square Well
96-well plate, Transparent, -, 2,0 ml
Catalog # 43001-0020
   Low Manufacturing Impact
Supplier:   Avantor
J.T.Baker®, Plates, Square Well
Catalog # 43001-0020
Supplier:  Avantor
   Low Manufacturing Impact
J.T.Baker®, Plates, Square Well
J.T.Baker®, Plates, Square Well
96-well plate, Transparent, -, 2,0 ml
Price
92,84 €
Log in for availability
1 * 20 items
Product Details & Documents
These deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
  • Chemical resistant
  • V-bottom shape for 24- and 48-wells maximises liquid removal and aids resuspension
  • U-bottom shape for 96 wells
  • No inner edges to interfere with magnetic bead collection
  • Raised well rims to improve heat sealing
  • Conical base aids sample concentration, reconstitution and centrifugation
For a maximum capacity utilisation.
Specifications
  • Sterile:
    -
  • Colour:
    Transparent
  • Description:
    96-well plate
  • Well volume:
    2,0 ml
  • Cat. No.:
    43001-0020
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Optical property
    Material
    Availability
    Price
  • Capmat for J.T.Baker plate square well 96
    Capmat for J.T.Baker plate square well 96
    43001-0022
    Transparent
    EVA
    Log in for availability
    1 * 20 items - 49,13 €
Product Information
  • Catalog #
    Optical property
    Material
    Availability
    Price
  • Capmat for J.T.Baker plate square well 48
    Capmat for J.T.Baker plate square well 48
    43001-0069
    Transparent
    EVA
    Log in for availability
    1 * 20 items - 57,88 €
Product Information
  • Catalog #
    Optical property
    Material
    Availability
    Price
  • Capmat for J.T.Baker plate square well 24
    Capmat for J.T.Baker plate square well 24
    43001-0070
    Transparent
    EVA
    Log in for availability
    1 * 20 items - 63,65 €