J.T.Baker®, Tip Box for Tecan Instruments
Tip box for C-Tray 50 and 200 µl tips
Catalog # 45009-3000
Supplier: Avantor
J.T.Baker®, Tip Box for Tecan Instruments
Catalog # 45009-3000
Supplier: Avantor
Jump to:
Product Details & Documents
The J.T.Baker® Tip Boxes are especially made as an accessory for pipette tips Type Tecan with C-Tray.
- Two different sizes available
Compatible tips are not included but need to be ordered separately.
Documents
Avantor J.T.Baker Robotic Consumables
Product Literature
Specifications
- Description:Tip box for C-Tray 50 and 200 µl tips
- Cat. No.:45009-3000
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact