Orders LinkContactUsLinkComponent
J.T.Baker®, Tip Box for Tecan Instruments
J.T.Baker®, Tip Box for Tecan Instruments
Tip box for C-Tray 1000 µl tips
Catalog # 45009-2000
   Low Manufacturing Impact
Supplier:   Avantor
J.T.Baker®, Tip Box for Tecan Instruments
Catalog # 45009-2000
Supplier:  Avantor
   Low Manufacturing Impact
J.T.Baker®, Tip Box for Tecan Instruments
J.T.Baker®, Tip Box for Tecan Instruments
Tip box for C-Tray 1000 µl tips
Price
16,59 €
Log in for availability
1 * 5 items
Product Details & Documents
The J.T.Baker® Tip Boxes are especially made as an accessory for pipette tips Type Tecan with C-Tray.
  • Two different sizes available
Compatible tips are not included but need to be ordered separately.
Documents
Specifications
  • Description:
    Tip box for C-Tray 1000 µl tips
  • Cat. No.:
    45009-2000
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Environmentally Preferable:
    Low Manufacturing Impact