Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Chloroparaffin C10-C13 51,5% Cl 100 µg/Ml In Cyclohexane 1 * 1 mL
Catalog # EHERA23105100CY
Supplier:
EHRENSTORFER
Expand image
Chloroparaffin C10-C13 51,5% Cl 100 µg/Ml In Cyclohexane 1 * 1 mL
Catalog # EHERA23105100CY
Supplier:
EHRENSTORFER