Order Entry
Export
Orders LinkContactUsLinkComponent
ToxinSensor™ single tests kit with standard

ToxinSensor™ single tests kit with standard

Sensitivity: 0,06 EU/ml, 40 assays, 1 KIT
Catalog # GENSL00857-40
Supplier:   Genscript
ToxinSensor™ single tests kit with standard
Catalog # GENSL00857-40
Supplier:  Genscript
CAS Number:  
ToxinSensor™ single tests kit with standard

Sensitivity: 0,06 EU/ml, 40 assays, 1 KIT

CAS Number:  
Price
335,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

ToxinSensor™ single tests kit with standard is designed to be a qualitative In Vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.

  • High reproducibility
  • Broad detection range
  • Easy to use
  • Includes one bottle of standard

Tachypleus Amebocyte Lysate (TAL) as supplied is to be reconstituted with sample or control directly. After incubation for 1 hour, and in the presence of endotoxin, gelation occurs; in the absence of endotoxin, gelation does not occur. GenScript supply a series of ToxinSensor™ Single Tests Kits with Standard with different sensitivity (0,03 EU/ml, 0,06 EU/ml, 0,125 EU/ml and 0,25 EU/ml). When using the kit, you just need to add 200 µl of sample or control to TAL, then, wait for the required incubation time to allow for gel to form. There is no need to reconstitute the system with TAL Reagent Water before use.

Documents

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Sensitivity: 0,06 EU/ml
  • For:
    40 assays
  • Cat. No.:
    GENSL00857-40
  • Sensitivity:
    0,06 EU/ml

Specifications