Order Entry
Export
Orders LinkContactUsLinkComponent
Cryogenic tubes, CryoTubes™, Nunc™

Cryogenic tubes, CryoTubes™, Nunc™

Cryogenic tubes with writing area, 4,50 ml, U, 12,5×92 mm, Yes, Clear
Catalog # 479-3202
Cryogenic tubes, CryoTubes™, Nunc™
Catalog # 479-3202
Cryogenic tubes, CryoTubes™, Nunc™
Cryogenic tubes, CryoTubes™, Nunc™

Cryogenic tubes with writing area, 4,50 ml, U, 12,5×92 mm, Yes, Clear

Price
951,00 €
Log in for availability
1 * 1.200 items
Product Details & Documents

PP tubes with PP screw cap (internal thread vials) or PE screw cap (external thread vials) and white writing area, sterile. Developed for storage of cell cultures, bacteria and other samples at extremely low temperatures. Suitable for use in freezers as well as in the vapour phase of liquid nitrogen*.

Non toxic, non pyrogenic.

Conform to IATA regulations for the air transportation of biological samples.

* Warning: Do not use CryoTubes™ in the liquid phase of liquid nitrogen unless correctly sealed in Nunc CryoFlex™ tubing. Improper use may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build up, resulting in possible explosion or biohazard release. The tubes 479-6816 and 479-6853 are suitable for storage to −80 °C only.

* without writing area
** free standing
*** starfoot

Specifications
  • Cap included:
    Yes
  • Ø×H:
    12,5×92 mm
  • Capacity:
    4,50 ml
  • Description:
    Cryogenic tubes with writing area
  • Seq:
    10
  • Cap colour:
    Clear
  • Base:
    U